1-(4-Hydroxyphenyl)piperazin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: p-(1-Piperazinyl)phenol

OR6840-100G OR6840-25G
APOSOR6840-100GEA 164 EUR
APOSOR6840-100G APOSOR6840-25G
1-(4-Hydroxyphenyl)piperazin 98%
1-(4-Hydroxyphenyl)piperazin
Formel: C₁₀H₁₄N₂O
Molecular Weight: 178,23 g/mol
Schmelzpunkt: 218…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066156
CAS-Nummer: 56621-48-8
EINECS: 260-289-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR