Ethylpiperazin-1-carboxylat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Ethoxycarbonyl piperazine, Ethyl N-piperazinecarboxylate
OR6852-500G
APOSOR6852-500GEA
185
EUR
APOSOR6852-500G
Ethylpiperazin-1-carboxylat 98%
Ethylpiperazin-1-carboxylat
Formel:
C₇H₁₄N₂O₂ Molecular Weight: 158,2 g/mol Siedepunkt: 272…274 °C Dichte: 1,08 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005964 CAS-Nummer: 120-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...