1-(4-Cyanphenyl)piperazin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Piperazin-1-yl)benzonitrile, 4-(1-Piperazinyl)benzonitrile, 1-(4-Cyanophenyl)piperazine

OR6866-5G OR6866-25G
APOSOR6866-5GEA 65.4 EUR
APOSOR6866-5G APOSOR6866-25G
1-(4-Cyanphenyl)piperazin 97%
1-(4-Cyanphenyl)piperazin
Formel: C₁₁H₁₃N₃
Molecular Weight: 187,2410 g/mol
Schmelzpunkt: 83…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320890
CAS-Nummer: 68104-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR