1-(2-Nitrophenyl)piperazin

Lieferant: Apollo Scientific
Achtung

OR6868-10G OR6868-1G OR6868-100G
APOSOR6868-10GEA 132 EUR
APOSOR6868-10G APOSOR6868-1G APOSOR6868-100G
1-(2-Nitrophenyl)piperazin
1-(2-Nitrophenyl)piperazin
Formel: C₁₀H₁₃N₃O₂
Siedepunkt: 142…144 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00040728
CAS-Nummer: 59084-06-9
EINECS: 261-593-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR