1-(4-Nitrophenyl)piperazin 97%

Lieferant: Apollo Scientific
Achtung

OR6870-100G OR6870-25G OR6870-5G
APOSOR6870-100GEA 104 EUR
APOSOR6870-100G APOSOR6870-25G APOSOR6870-5G
1-(4-Nitrophenyl)piperazin 97%
1-(4-Nitrophenyl)piperazin
Formel: C₁₀H₁₃N₃O₂
Schmelzpunkt: 129…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005961
CAS-Nummer: 6269-89-2
EINECS: 228-443-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR