4-Tolylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Methylphenyl)metaborsäure, 4-Methylphenylboronsäure, 4-Methylbenzolboronsäure, Dihydroxy(4-methylphenyl)boran

OR6888-25G OR6888-100G OR6888-5G
APOSOR6888-25GEA 24.8 EUR
APOSOR6888-25G APOSOR6888-100G APOSOR6888-5G
4-Tolylboronsäure
4-Tolylboronsäure
Formel: C₇H₉BO₂
Molecular Weight: 135.96 g/mol
Schmelzpunkt: 256…263 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039138
CAS-Nummer: 5720-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR