4-Chlor-6-nitro-o-toluidin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-2-methyl-6-nitroaniline

OR6914-250MG OR6914-5G
APOSOR6914-250MGEA 21.9 EUR
APOSOR6914-250MG APOSOR6914-5G
4-Chlor-6-nitro-o-toluidin 98%
4-Chlor-6-nitro-o-toluidin
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186,60 g/mol
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320687
CAS-Nummer: 62790-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR