Diisopropyl(2-piperazin-1-ylethyl)amine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-{2-[Bis(isopropyl)amino]ethyl}piperazine

OR6919-10G OR6919-2G OR6919-5G
APOSOR6919-10GEA 242 EUR
APOSOR6919-10G APOSOR6919-2G APOSOR6919-5G
Diisopropyl(2-piperazin-1-ylethyl)amine 97%
Diisopropyl(2-piperazin-1-ylethyl)amine
Formel: C₁₂H₂₇N₃
Molecular Weight: 213,37 g/mol
Siedepunkt: 64…66 °C/0,1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01320902
CAS-Nummer: 59955-93-0
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR