1-bromo-2,4-dimethyl-5-nitrobenzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2,4-dimethyl-5-nitrobenzene
OR6965-100G
OR6965-25G
OR6965-5G
APOSOR6965-100GEA
154
EUR
APOSOR6965-100G
APOSOR6965-25G
APOSOR6965-5G
1-bromo-2,4-dimethyl-5-nitrobenzene
1-bromo-2,4-dimethyl-5-nitrobenzene
Formel:
C₈H₈BrNO₂ Molecular Weight: 230,061 g/mol Schmelzpunkt: 55…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075824 CAS-Nummer: 69383-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...