2,3-Dihydro-5-(methylsulphonyl)-(1H)-indole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Methylsulphonyl)indoline

OR6977-1G
APOSOR6977-1GEA 60.9 EUR
APOSOR6977-1G
2,3-Dihydro-5-(methylsulphonyl)-(1H)-indole 95%
2,3-Dihydro-5-(methylsulphonyl)-(1H)-indole
Formel: C₉H₁₁NO₂S
Molecular Weight: 197,26 g/mol
Schmelzpunkt: 129…131 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01319203
CAS-Nummer: 387350-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR