4-(4-Chlorophenyl)thiophene-2-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR6996-1G OR6996-5G OR6996-10G
APOSOR6996-1GEA 229 EUR
APOSOR6996-1G APOSOR6996-5G APOSOR6996-10G
4-(4-Chlorophenyl)thiophene-2-carboxylic acid 95+%
4-(4-Chlorophenyl)thiophene-2-carboxylic acid
Formel: C₁₁H₇ClO₂S
Molecular Weight: 238.69 g/mol
Schmelzpunkt: 232…235 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01319431
CAS-Nummer: 386715-46-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR