4-Formylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Formylphenyl)metaborsäure, 4-Boronobenzaldehyd, Dihydroxy(4-formylphenyl)boran, 4-Formylbenzolboronsäure

OR6999-100G OR6999-25G OR6999-5G
APOSOR6999-100GEA 109 EUR
APOSOR6999-100G APOSOR6999-25G APOSOR6999-5G
4-Formylphenylboronsäure
4-Formylphenylboronsäure
Formel: C₇H₇BO₃
Molecular Weight: 149,94 g/mol
Schmelzpunkt: 237…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151823
CAS-Nummer: 87199-17-5
EINECS: 438-670-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR