1-Boc-4-(hydroxymethyl)piperidin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-4-piperidinemethanol, N-BOC-4-Piperidinemethanol
OR7003-100G
OR7003-25G
OR7003-5G
APOSOR7003-100GEA
48
EUR
APOSOR7003-100G
APOSOR7003-25G
APOSOR7003-5G
1-Boc-4-(hydroxymethyl)piperidin 97%
1-Boc-4-(hydroxymethyl)piperidin
Formel:
C₁₁H₂₁NO₃ Molecular Weight: 215.29 g/mol Schmelzpunkt: 78…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02094488 CAS-Nummer: 123855-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...