2-Methyl-3-phenylbenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-[1,1'-biphenyl]-3-carboxylic acid

OR70044-1G OR70044-5G
APOSOR70044-1GEA 174 EUR
APOSOR70044-1G APOSOR70044-5G
2-Methyl-3-phenylbenzoesäure 98%
2-Methyl-3-phenylbenzoic acid
Formel: C₁₄H₁₂O₂
Schmelzpunkt: 156…160 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD11554177
CAS-Nummer: 115363-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR