3,4-Dichlorbenzophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: (3,4-Dichlorophenyl)(phenyl)methanone

OR70083-100G OR70083-25G
APOSOR70083-100GEA 87 EUR
APOSOR70083-100G APOSOR70083-25G
3,4-Dichlorbenzophenon
3,4-Dichlorbenzophenon
Formel: C₁₃H₈Cl₂O
Molecular Weight: 251,11 g/mol
Schmelzpunkt: 100…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000552
CAS-Nummer: 6284-79-3
EINECS: 228-509-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR