2-Chlor-9,10-anthrachinon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloranthrachinon
OR70084-50G
OR70084-25G
APOSOR70084-50GEA
0
EUR
APOSOR70084-50G
APOSOR70084-25G
2-Chlor-9,10-anthrachinon
2-Chlor-9,10-anthrachinon
Formel:
C₁₄H₇ClO₂ Molecular Weight: 242,66 g/mol Siedepunkt: 292 °C/30mmHg Schmelzpunkt: 207…212 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001227 CAS-Nummer: 131-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...