Tris(4-bromophenyl)amine 98%

Lieferant: Apollo Scientific
Achtung

OR70090-100G OR70090-25G OR70090-5G
APOSOR70090-100GEA 109 EUR
APOSOR70090-100G APOSOR70090-25G APOSOR70090-5G
Tris(4-bromophenyl)amine 98%
Tris(4-bromophenyl)amine
Formel: C₁₈H₁₂Br₃N
Molecular Weight: 482.01 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009665
CAS-Nummer: 4316-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR