2-Brom-3-iodtoluol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-1-iodo-3-methylbenzene
OR70218-5G
APOSOR70218-5GEA
59.6
EUR
APOSOR70218-5G
2-Brom-3-iodtoluol 95%
2-Brom-3-iodtoluol
Formel:
C₇H₆BrI Molecular Weight: 296,9310 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11977389 CAS-Nummer: 888214-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...