Methyl-2,3-dimethylbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2,3-dimethylbenzoate

OR7024-10G OR7024-1G OR7024-5G
APOSOR7024-10GEA 74.1 EUR
APOSOR7024-10G APOSOR7024-1G APOSOR7024-5G
Methyl-2,3-dimethylbenzoat
Methyl-2,3-dimethylbenzoat
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2000 g/mol
Siedepunkt: 104…105 °C/12mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD06203773
CAS-Nummer: 15012-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR