3-Amino-2,4-dimethylpyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR7029-1G
OR7029-250MG
OR7029-5G
APOSOR7029-1GEA
101
EUR
APOSOR7029-1G
APOSOR7029-250MG
APOSOR7029-5G
3-Amino-2,4-dimethylpyridin 98%
3-Amino-2,4-dimethylpyridine
Formel:
C₇H₁₀N₂ Molecular Weight: 122,17 g/mol Schmelzpunkt: 37…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08235192 CAS-Nummer: 1073-21-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...