3,5-Diamino-1H-1,2,4-triazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Guanazol
OR7056-100G
OR7056-25G
OR7056-10G
APOSOR7056-100GEA
60.9
EUR
APOSOR7056-100G
APOSOR7056-25G
APOSOR7056-10G
3,5-Diamino-1H-1,2,4-triazol
3,5-Diamino-1H-1,2,4-triazol
Formel:
C₂H₅N₅ Molecular Weight: 99.1 g/mol Siedepunkt: 425 Schmelzpunkt: 202…205 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005233 CAS-Nummer: 1455-77-2 EINECS: 215-937-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...