3-Methyl-4-isoxazolcarbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methylisoxazole-4-carboxylic acid

OR7097-1G OR7097-250MG OR7097-5G
APOSOR7097-1GEA 34.8 EUR
APOSOR7097-1G APOSOR7097-250MG APOSOR7097-5G
3-Methyl-4-isoxazolcarbonsäure
3-Methyl-4-isoxazolecarboxylic acid
Formel: C₅H₅NO₃
Molecular Weight: 127,10 g/mol
Schmelzpunkt: 183…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318161
CAS-Nummer: 17153-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR