[1,2,4]Triazol-1-ylacetic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1H-1,2,4-Triazol-1-yl)acetic acid
OR7103-1G
OR7103-5G
APOSOR7103-1GEA
106
EUR
APOSOR7103-1G
APOSOR7103-5G
[1,2,4]Triazol-1-ylacetic acid 98%
[1,2,4]Triazol-1-ylacetic acid
Formel:
C₄H₅N₃O₂ Molecular Weight: 127.1 g/mol Schmelzpunkt: 201…205 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036051 CAS-Nummer: 28711-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...