1-Benzyl-4-cyclopropylaminopiperidine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7115-1G
APOSOR7115-1GEA
59.6
EUR
APOSOR7115-1G
1-Benzyl-4-cyclopropylaminopiperidine 95+%
1-Benzyl-4-cyclopropylaminopiperidine
Formel:
C₁₅H₂₂N₂ Siedepunkt: 118 °C/0,01mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD01308091 CAS-Nummer: 387358-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...