3'-Chlor-2'-methylacetanilid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR7119-100G
OR7119-25G
APOSOR7119-100GEA
279
EUR
APOSOR7119-100G
APOSOR7119-25G
3'-Chlor-2'-methylacetanilid 97%
3'-Chlor-2'-methylacetanilid
Formel:
C₉H₁₀ClNO Molecular Weight: 183,64 g/mol Schmelzpunkt: 155…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00018264 CAS-Nummer: 7463-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...