6-Hydroxy-3-coumaranon 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Hydroxybenzofuran-3(2H)-one, 6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one

OR7143-5G OR7143-25G OR7143-100G
APOSOR7143-5GEA 0 EUR
APOSOR7143-5G APOSOR7143-25G APOSOR7143-100G
6-Hydroxy-3-coumaranon 95%
6-Hydroxy-3-coumaranon
Formel: C₈H₆O₃
Molecular Weight: 150.14 g/mol
Schmelzpunkt: 242…246 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068174
CAS-Nummer: 6272-26-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR