5-Acetyl-2-thiophenboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Acetylthien-5-yl)boronic acid,(5-Acetylthien-2-yl)boronic acid,5-Acetyl-2-thiopheneboronic acid,5-Acetylthiophen-2-ylboronic acid, 5-Acetyl-2-thienylboronic acid, 5-Acetylthiophene-2-boronic acid, 5-Acetyl-2-thiopheneboronic acid

OR7147-25G OR7147-5G OR7147-100G
APOSOR7147-25GEA 92.8 EUR
APOSOR7147-25G APOSOR7147-5G APOSOR7147-100G
5-Acetyl-2-thiophenboronsäure
5-Acetyl-2-thiophenboronsäure
Formel: C₆H₇BO₃S
Molecular Weight: 170,00 g/mol
Schmelzpunkt: 142…148 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01075681
CAS-Nummer: 206551-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR