Benzo[b]furan-5-boronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzofuran-5-ylboronic acid
OR7149-1G
OR7149-5G
APOSOR7149-1GEA
85.7
EUR
APOSOR7149-1G
APOSOR7149-5G
Benzo[b]furan-5-boronsäure
Benzo[b]furan-5-boronsäure
Formel:
C₈H₇BO₃ Molecular Weight: 161,9500 g/mol Schmelzpunkt: 216…220 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04038999 CAS-Nummer: 331834-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...