2-Brom-5-pyridinboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromopyridine-5-boronic acid pinacol ester, 2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

OR7165-250MG OR7165-5G OR7165-1G
APOSOR7165-250MGEA 21.9 EUR
APOSOR7165-250MG APOSOR7165-5G APOSOR7165-1G
2-Brom-5-pyridinboronsäurepinakolester
2-Brom-5-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BBrNO₂
Molecular Weight: 283.96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412794
CAS-Nummer: 214360-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR