2-(tert-Butoxymethyl)phenylboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(tert-Butoxymethyl)benzeneboronic acid
OR7173-25G
OR7173-5G
OR7173-1G
APOSOR7173-25GEA
1520
EUR
APOSOR7173-25G
APOSOR7173-5G
APOSOR7173-1G
2-(tert-Butoxymethyl)phenylboronic acid
2-(tert-Butoxymethyl)phenylboronic acid
Formel:
C₁₁H₁₇BO₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD03425930 CAS-Nummer: 373384-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...