4-Butoxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Butoxyphenyl)boronic acid, 4-n-Butoxyphenylboronic acid
OR7178-1G
OR7178-5G
OR7178-10G
APOSOR7178-1GEA
21.9
EUR
APOSOR7178-1G
APOSOR7178-5G
APOSOR7178-10G
4-Butoxyphenylboronsäure
4-Butoxyphenylboronsäure
Formel:
C₁₀H₁₅BO₃ Molecular Weight: 194,04 g/mol Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427054 CAS-Nummer: 105365-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...