3-Chlor-4-pyridinboronsäure 97%

Lieferant: Apollo Scientific
Achtung

OR7185-5G OR7185-1G OR7185-250MG OR7185-10G
APOSOR7185-5GEA 87 EUR
APOSOR7185-5G APOSOR7185-1G APOSOR7185-250MG APOSOR7185-10G
3-Chlor-4-pyridinboronsäure 97%
3-Chloro-4-pyridineboronic acid
Formel: C₅H₅BClNO₂
Molecular Weight: 157.36 g/mol
Schmelzpunkt: 95…97 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD03425942
CAS-Nummer: 458532-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR