2-Chlorchinolin-3-boronsäure 95%

Lieferant: Apollo Scientific
Achtung

OR7186-1G OR7186-5G
APOSOR7186-1GEA 29 EUR
APOSOR7186-1G APOSOR7186-5G
2-Chlorchinolin-3-boronsäure 95%
2-Chlorchinolin-3-boronsäure
Formel: C₉H₇BClNO₂
Molecular Weight: 207,4214 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01114675
CAS-Nummer: 128676-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR