4-Cyclohexylphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyclohexylbenzeneboronic acid, (4-Cyclohexylphenyl)boronic acid

OR7189-1G OR7189-5G
APOSOR7189-1GEA 219 EUR
APOSOR7189-1G APOSOR7189-5G
4-Cyclohexylphenylboronsäure 98%
4-Cyclohexylphenylboronsäure
Formel: C₁₂H₁₇BO₂
Molecular Weight: 204.08 g/mol
Schmelzpunkt: 220 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02093068
CAS-Nummer: 374538-04-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR