2-(Cyclopropylmethoxy)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

OR7192-1G OR7192-5G
APOSOR7192-1GEA 350 EUR
APOSOR7192-1G APOSOR7192-5G
2-(Cyclopropylmethoxy)phenylboronsäure
2-(Cyclopropylmethoxy)phenylboronsäure
Formel: C₁₀H₁₃BO₃
Lagertemperatur: Kühlschrank
MDL: MFCD04039015
CAS-Nummer: 1050510-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR