4-(N,N-Dimethylamino)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Dimethylamino)benzeneboronic acid
OR7205-1G
OR7205-5G
OR7205-25G
APOSOR7205-1GEA
72.5
EUR
APOSOR7205-1G
APOSOR7205-5G
APOSOR7205-25G
4-(N,N-Dimethylamino)phenylboronsäure 98%
4-(N,N-Dimethylamino)phenylboronsäure
Formel:
C₈H₁₂BNO₂ Molecular Weight: 165,00 g/mol Schmelzpunkt: 227 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01074642 CAS-Nummer: 28611-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...