3-(Methoxymethyl)phenylboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methoxymethylphenylboronic acid, (3-(Methoxymethyl)phenyl)boronic acid
OR7232-1G
OR7232-250MG
OR7232-5G
APOSOR7232-1GEA
150
EUR
APOSOR7232-1G
APOSOR7232-250MG
APOSOR7232-5G
3-(Methoxymethyl)phenylboronic acid
3-(Methoxymethyl)phenylboronic acid
Formel:
C₈H₁₁BO₃ Molecular Weight: 165,99 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03412069 CAS-Nummer: 142273-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...