4-(Methoxymethyl)benzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Methoxymethyl)phenylboronic acid, (4-(Methoxymethyl)phenyl)boronic acid

OR7233-25G OR7233-5G
APOSOR7233-25GEA 169 EUR
APOSOR7233-25G APOSOR7233-5G
4-(Methoxymethyl)benzeneboronic acid 97%
4-(Methoxymethyl)benzeneboronic acid
Formel: C₈H₁₁BO₃
Molecular Weight: 165.983 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788426
CAS-Nummer: 279262-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR