2-(Morpholinomethyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-(Morpholinomethyl)phenyl)boronic acid
OR7241-1G
OR7241-5G
APOSOR7241-1GEA
112
EUR
APOSOR7241-1G
APOSOR7241-5G
2-(Morpholinomethyl)phenylboronsäure 98%
2-(Morpholinomethyl)phenylboronsäure
Formel:
C₁₁H₁₆BNO₃ Molecular Weight: 221.0600 g/mol Schmelzpunkt: 222…226 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01319041 CAS-Nummer: 223433-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...