(-)-Cotinin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Cotinine, (S)-1-Methyl-5-(pyridin-3-yl)pyrrolidin-2-one
OR7251T-1G
OR7251T-250MG
APOSOR7251T-1GEA
859
EUR
APOSOR7251T-1G
APOSOR7251T-250MG
(-)-Cotinin
(-)-Cotinin
A major metabolite of nicotine in humans.
Formel:
C₁₀H₁₂N₂O Molecular Weight: 176.22 g/mol Siedepunkt: 145…150 °C/3mmHg Schmelzpunkt: 48…50 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077696 CAS-Nummer: 486-56-6 EINECS: 207-634-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...