Pyridine-3-boronic acid neopentylglycol ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine

OR7259-10G OR7259-1G OR7259-5G
APOSOR7259-10GEA 161 EUR
APOSOR7259-10G APOSOR7259-1G APOSOR7259-5G
Pyridine-3-boronic acid neopentylglycol ester 97%
Pyridine-3-boronic acid neopentylglycol ester
Formel: C₁₀H₁₄BNO₂
Molecular Weight: 191,04 g/mol
Schmelzpunkt: 91…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425979
CAS-Nummer: 845885-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR