5-Pyrimidinboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR7262-1G
OR7262-5G
OR7262-10G
APOSOR7262-1GEA
21.9
EUR
APOSOR7262-1G
APOSOR7262-5G
APOSOR7262-10G
5-Pyrimidinboronsäure
5-Pyrimidineboronic acid
Formel:
C₄H₅BN₂O₂ Schmelzpunkt: 110…114 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03002366 CAS-Nummer: 109299-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...