4'-Acetylbiphenyl-4-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4'-Acetyl-[1,1'-biphenyl]-4-carboxylic acid
OR7314-1G
OR7314-250MG
OR7314-5G
APOSOR7314-1GEA
46.4
EUR
APOSOR7314-1G
APOSOR7314-250MG
APOSOR7314-5G
4'-Acetylbiphenyl-4-carbonsäure
4'-Acetylbiphenyl-4-carbonsäure
Formel:
C₁₅H₁₂O₃ Molecular Weight: 240,2500 g/mol Schmelzpunkt: 281…284 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00230828 CAS-Nummer: 114691-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...