2-Formyl-4-methoxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR7336-1G
OR7336-5G
OR7336-25G
APOSOR7336-1GEA
23.2
EUR
APOSOR7336-1G
APOSOR7336-5G
APOSOR7336-25G
2-Formyl-4-methoxyphenylboronsäure
2-Formyl-4-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ Schmelzpunkt: 166…170 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01319043 CAS-Nummer: 139962-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...