3'-Chlorbiphenyl-3-carbonsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
OR7362-1G
OR7362-5G
APOSOR7362-1GEA
125
EUR
APOSOR7362-1G
APOSOR7362-5G
3'-Chlorbiphenyl-3-carbonsäure 96%
3'-Chlorbiphenyl-3-carbonsäure
Formel:
C₁₃H₉ClO₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD03424595 CAS-Nummer: 168619-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...