4'-Chlorbiphenyl-2-carbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4'-Chloro-[1,1'-biphenyl]-2-carboxylic acid, 4'-Chloro[1,1'-biphenyl]-2-carboxylic acid
OR7364-1G
OR7364-5G
APOSOR7364-1GEA
170
EUR
APOSOR7364-1G
APOSOR7364-5G
4'-Chlorbiphenyl-2-carbonsäure 98%
4'-Chlorbiphenyl-2-carbonsäure
Formel:
C₁₃H₉ClO₂ Molecular Weight: 232.67 g/mol Schmelzpunkt: 167…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426504 CAS-Nummer: 7079-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...