2'-Formylbiphenyl-2-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR7383-1G
OR7383-250MG
APOSOR7383-1GEA
265
EUR
APOSOR7383-1G
APOSOR7383-250MG
2'-Formylbiphenyl-2-carbonsäure
2'-Formylbiphenyl-2-carbonsäure
Formel:
C₁₄H₁₀O₃ Schmelzpunkt: 114…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039066 CAS-Nummer: 6720-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...