3'-Formylbiphenyl-4-carbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3'-Formyl-[1,1'-biphenyl]-4-carboxylic acid

OR7388-100MG OR7388-1G OR7388-5G
APOSOR7388-100MGEA 21.9 EUR
APOSOR7388-100MG APOSOR7388-1G APOSOR7388-5G
3'-Formylbiphenyl-4-carbonsäure
3'-Formylbiphenyl-4-carbonsäure
Formel: C₁₄H₁₀O₃
Schmelzpunkt: 231…233 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039069
CAS-Nummer: 222180-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR