3-Brom-6-chlorimidazo[1,2-b]pyridazin 98%

Lieferant: Apollo Scientific
Achtung

OR7413-1G OR7413-250MG OR7413-10G OR7413-25G
APOSOR7413-1GEA 0 EUR
APOSOR7413-1G APOSOR7413-250MG APOSOR7413-10G APOSOR7413-25G
3-Brom-6-chlorimidazo[1,2-b]pyridazin 98%
3-Brom-6-chlorimidazo[1,2-b]pyridazin
Formel: C₆H₃BrClN₃
Molecular Weight: 232,47 g/mol
Schmelzpunkt: 122…126 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09027274
CAS-Nummer: 13526-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR