3-(2-Bromphenylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
OR7418-1G
OR7418-5G
APOSOR7418-1GEA
125
EUR
APOSOR7418-1G
APOSOR7418-5G
3-(2-Bromphenylcarbamoyl)phenylboronsäure 98%
3-(2-Bromphenylcarbamoyl)phenylboronsäure
Formel:
C₁₃H₁₁BBrNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027206 CAS-Nummer: 874288-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...